4-(3-Hydroxypropyl)benzonitrile 95%

Lieferant: Apollo Scientific

OR931934-1G
APOSOR931934-1GEA 151 EUR
APOSOR931934-1G
4-(3-Hydroxypropyl)benzonitrile 95%
4-(3-Hydroxypropyl)benzonitrile
Formel: C₁₀H₁₁NO
Molecular Weight: 161.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09028771
CAS-Nummer: 83101-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR