Chroman-4-amine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dihydro-2H-1-benzopyran-4-amine

OR931982-1G
APOSOR931982-1GEA 102 EUR
APOSOR931982-1G
Chroman-4-amine 97%
Chroman-4-amine
Formel: C₉H₁₁NO
Molecular Weight: 149,1897 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00730969
CAS-Nummer: 53981-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR