(S)-tert-Butyl (2-aminopropyl)carbamate 97%

Lieferant: Apollo Scientific

OR932018-1G
APOSOR932018-1GEA 272 EUR
APOSOR932018-1G
(S)-tert-Butyl (2-aminopropyl)carbamate 97%
(S)-tert-Butyl (2-aminopropyl)carbamate
Formel: C₈H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11112235
CAS-Nummer: 121103-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR