3,5-Dichloro-4-(chloromethyl)pyridine 98%
Lieferant: Apollo Scientific
OR932052-1G
APOSOR932052-1GEA
734
EUR
APOSOR932052-1G
3,5-Dichloro-4-(chloromethyl)pyridine 98%
3,5-Dichloro-4-(chloromethyl)pyridine
Formel:
C₆H₄Cl₃N Molecular Weight: 196,462 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14582993 CAS-Nummer: 264123-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...