3,4-Dihydro-1H-2-benzopyran-3-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: Isochroman-3-carboxylic acid

OR932058-1G
APOSOR932058-1GEA 87 EUR
APOSOR932058-1G
3,4-Dihydro-1H-2-benzopyran-3-carboxylic acid 95%
3,4-Dihydro-1H-2-benzopyran-3-carboxylic acid
Formel: C₁₀H₁₀O₃
Molecular Weight: 178,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18416383
CAS-Nummer: 1261578-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR