4-Chloro-n,n-dimethylpyridin-2-amine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-N,N-dimethylpyridin-2-amine

OR932120-1G
APOSOR932120-1GEA 200 EUR
APOSOR932120-1G
4-Chloro-n,n-dimethylpyridin-2-amine 95%
4-Chloro-n,n-dimethylpyridin-2-amine
Formel: C₇H₉ClN₂
Molecular Weight: 156,613 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09031036
CAS-Nummer: 735255-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR