Quinolin-4-ylmethanol 95%

Lieferant: Apollo Scientific

Synonyme: 4-(Hydroxymethyl)quinoline

OR932181-1G
APOSOR932181-1GEA 149 EUR
APOSOR932181-1G
Quinolin-4-ylmethanol 95%
Quinolin-4-ylmethanol
Formel: C₁₀H₉NO
Molecular Weight: 159,185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01006059
CAS-Nummer: 6281-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR