3-Cyano-benzenepropanoic acid 95%

Lieferant: Apollo Scientific

Synonyme: 3-Cyanobenzenepropanoic acid

OR932222-1G
APOSOR932222-1GEA 97.3 EUR
APOSOR932222-1G
3-Cyano-benzenepropanoic acid 95%
3-Cyano-benzenepropanoic acid
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061831
CAS-Nummer: 42287-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR