3-Methyl-2-cyclohexen-1-on 98%
Lieferant: Apollo Scientific
Synonyme:
3-Methylcyclohex-2-enon
OR932249-100G
APOSOR932249-100GEA
154
EUR
APOSOR932249-100G
3-Methyl-2-cyclohexen-1-on 98%
3-Methyl-2-cyclohexen-1-on
Formel:
C₇H₁₀O Molecular Weight: 110.16 g/mol Siedepunkt: 197 °C Dichte: 0.97 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001581 CAS-Nummer: 1193-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...