3-Chlor-5-methylbenzonitril 97%

Lieferant: Apollo Scientific

OR932275-5G
APOSOR932275-5GEA 72.5 EUR
APOSOR932275-5G
3-Chlor-5-methylbenzonitril 97%
3-Chlor-5-methylbenzonitril
Formel: C₈H₆ClN
Molecular Weight: 151.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800704
CAS-Nummer: 189161-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR