3-Chromanecarboxylic acid 95%

Lieferant: Apollo Scientific

OR932278-1G
APOSOR932278-1GEA 58 EUR
APOSOR932278-1G
3-Chromanecarboxylic acid 95%
3-Chromanecarboxylic acid
Formel: C₁₀H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03783554
CAS-Nummer: 115822-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR