5-(3-Chlorophenyl)-3,4-dihydro-2H-pyrrole 95%

Lieferant: Apollo Scientific

OR932281-1G
APOSOR932281-1GEA 464 EUR
APOSOR932281-1G
5-(3-Chlorophenyl)-3,4-dihydro-2H-pyrrole 95%
5-(3-Chlorophenyl)-3,4-dihydro-2H-pyrrole
Formel: C₁₀H₁₀ClN
Molecular Weight: 179,646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037876
CAS-Nummer: 374588-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR