6-Bromoquinazolin-4-amine 98%

Lieferant: Apollo Scientific
Achtung

OR932284-5G
APOSOR932284-5GEA 130 EUR
APOSOR932284-5G
6-Bromoquinazolin-4-amine 98%
6-Bromoquinazolin-4-amine
Formel: C₈H₆BrN₃
Molecular Weight: 224.0600 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02228849
CAS-Nummer: 21419-48-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR