C-[1,4]Dioxan-2-ylmethylamine 95%

Lieferant: Apollo Scientific

OR932330-250MG
APOSOR932330-250MGEA 484 EUR
APOSOR932330-250MG
C-[1,4]Dioxan-2-ylmethylamine 95%
C-[1,4]Dioxan-2-ylmethylamine
Formel: C₅H₁₁NO₂
Molecular Weight: 117,15 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01994527
CAS-Nummer: 88277-83-2
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR