2-Methylaminoethyl chloride hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-N-methylethanamine hydrochloride

OR932350-10G OR932350-1G OR932350-25G OR932350-100G
APOSOR932350-10GEA 68.3 EUR
APOSOR932350-10G APOSOR932350-1G APOSOR932350-25G APOSOR932350-100G
2-Methylaminoethyl chloride hydrochloride 97%
2-Methylaminoethyl chloride hydrochloride
Formel: C₃H₈ClN·ClH
Molecular Weight: 130.016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050517
CAS-Nummer: 4535-90-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR