5-Hydroxy-2-methylbenzaldehyde 95%

Lieferant: Apollo Scientific

OR932375-1G
APOSOR932375-1GEA 249 EUR
APOSOR932375-1G
5-Hydroxy-2-methylbenzaldehyde 95%
5-Hydroxy-2-methylbenzaldehyde
Formel: C₈H₈O₂
Molecular Weight: 136,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16997527
CAS-Nummer: 23942-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR