(R,R)-(-)-3,6-Octandiol 96%
Lieferant: Apollo Scientific
OR932401-500MG
APOSOR932401-500MGEA
667
EUR
APOSOR932401-500MG
(R,R)-(-)-3,6-Octandiol 96%
(R,R)-(-)-3,6-Octandiol
Formel:
C₈H₁₈O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631087 CAS-Nummer: 129619-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...