N, O-Bis (Trimethylsilyl)Hydroxylamin 95%

Lieferant: Apollo Scientific

OR932413-1G OR932413-5G
APOSOR932413-1GEA 0 EUR
APOSOR932413-1G APOSOR932413-5G
N, O-Bis (Trimethylsilyl)Hydroxylamin 95%
N, O-Bis (Trimethylsilyl)Hydroxylamin
Formel: C₆H₁₉NOSi₂
Molecular Weight: 177.39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008266
CAS-Nummer: 22737-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR