5,6-Dichloropyridazin-3(2H)-one 95%
Lieferant: Apollo Scientific
OR932432-5G
APOSOR932432-5GEA
69.6
EUR
APOSOR932432-5G
5,6-Dichloropyridazin-3(2H)-one 95%
5,6-Dichloropyridazin-3(2H)-one
Formel:
C₄H₂Cl₂N₂O Molecular Weight: 164.977 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04971420 CAS-Nummer: 17285-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...