Methyl-3-oxotetrahydrothiophene-2-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-oxotetrahydrothiophene-2-carboxylate

OR932444-1G
APOSOR932444-1GEA 78.3 EUR
APOSOR932444-1G
Methyl-3-oxotetrahydrothiophene-2-carboxylate 95%
Methyl-3-oxotetrahydrothiophene-2-carboxylate
Formel: C₆H₈O₃S
Molecular Weight: 160,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156156
CAS-Nummer: 2689-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR