(4-Chloro-5-methyl-pyrimidin-2-yl)-cyclopropyl-amine 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-N-cyclopropyl-5-methylpyrimidin-2-amine

OR932486-1G
APOSOR932486-1GEA 516 EUR
APOSOR932486-1G
(4-Chloro-5-methyl-pyrimidin-2-yl)-cyclopropyl-amine 95%
(4-Chloro-5-methyl-pyrimidin-2-yl)-cyclopropyl-amine
Formel: C₈H₁₀ClN₃
Molecular Weight: 183.638 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837194
CAS-Nummer: 1289385-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR