3-(3-Formyl-1H-indol-1-yl)propanenitrile 95%

Lieferant: Apollo Scientific

OR932512-1G
APOSOR932512-1GEA 435 EUR
APOSOR932512-1G
3-(3-Formyl-1H-indol-1-yl)propanenitrile 95%
3-(3-Formyl-1H-indol-1-yl)propanenitrile
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00484155
CAS-Nummer: 18109-11-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR