(R)-Methyl 4-(1-aminoethyl)benzoate 95%

Lieferant: Apollo Scientific

OR932545-5G
APOSOR932545-5GEA 212 EUR
APOSOR932545-5G
(R)-Methyl 4-(1-aminoethyl)benzoate 95%
(R)-Methyl 4-(1-aminoethyl)benzoate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06762071
CAS-Nummer: 912342-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR