4-Methyloxazole

Lieferant: Apollo Scientific

OR932677-5G
APOSOR932677-5GEA 56.7 EUR
APOSOR932677-5G
4-Methyloxazole
4-Methyloxazole
Formel: C₄H₅NO
Molecular Weight: 83.089 g/mol
Siedepunkt: 86…88 °C
Lagertemperatur: Kühlschrank
MDL: MFCD11110237
CAS-Nummer: 693-93-6
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR