1-(3-Nitrophenyl)-1H-pyrazole 95%

Lieferant: Apollo Scientific

OR932686-1G OR932686-250MG
APOSOR932686-1GEA 435 EUR
APOSOR932686-1G APOSOR932686-250MG
1-(3-Nitrophenyl)-1H-pyrazole 95%
1-(3-Nitrophenyl)-1H-pyrazole
Formel: C₉H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12192642
CAS-Nummer: 25688-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR