Methyl (R)-4-chloro-3-hydroxybutyrate 97%

Lieferant: Apollo Scientific

Synonyme: (R)-4-Chloro-3-hydroxybutyric Acid Methyl Ester

OR932699-5G
APOSOR932699-5GEA 0 EUR
APOSOR932699-5G
Methyl (R)-4-chloro-3-hydroxybutyrate 97%
Methyl (R)-4-chloro-3-hydroxybutyrate
Formel: C₅H₉ClO₃
Molecular Weight: 152,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00273367
CAS-Nummer: 88496-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR