2-Methyl-3-pyridinboronsäure Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methylpyridine-3-boronic acid hydrochloride, (2-Methylpyridin-3-yl)boronic acid hydrochloride

OR9327-1G
APOSOR9327-1GEA 321 EUR
APOSOR9327-1G
2-Methyl-3-pyridinboronsäure Hydrochlorid
2-Methyl-3-pyridinboronsäure Hydrochlorid
Formel: C₆H₉BClNO₂
Molecular Weight: 173,4100 g/mol
Schmelzpunkt: 181…186 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659489
CAS-Nummer: 1072952-34-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR