4,7-Dichloro-1,6-naphthyridine 96%

Lieferant: Apollo Scientific

OR932711-1G OR932711-250MG
APOSOR932711-1GEA 467 EUR
APOSOR932711-1G APOSOR932711-250MG
4,7-Dichloro-1,6-naphthyridine 96%
4,7-Dichloro-1,6-naphthyridine
Formel: C₈H₄Cl₂N₂
Molecular Weight: 199.037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044711
CAS-Nummer: 952138-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR