4-(2-Methoxyethyl)phenol 97%

Lieferant: Apollo Scientific

OR932723-100G OR932723-25G OR932723-500G
APOSOR932723-100GEA 25.8 EUR
APOSOR932723-100G APOSOR932723-25G APOSOR932723-500G
4-(2-Methoxyethyl)phenol 97%
4-(2-Methoxyethyl)phenol
Formel: C₉H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00017537
CAS-Nummer: 56718-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR