4-Chloro-7-methoxyquinazoline 95%

Lieferant: Apollo Scientific

OR932740-1G
APOSOR932740-1GEA 132 EUR
APOSOR932740-1G
4-Chloro-7-methoxyquinazoline 95%
4-Chloro-7-methoxyquinazoline
Formel: C₉H₇ClN₂O
Molecular Weight: 194.618 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738758
CAS-Nummer: 55496-52-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR