6-Chloro-2-ethyl-1H-benzimidazole 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-2-ethyl-1H-benzo[d]imidazole

OR932774-1G OR932774-250MG OR932774-5G
APOSOR932774-1GEA 115 EUR
APOSOR932774-1G APOSOR932774-250MG APOSOR932774-5G
6-Chloro-2-ethyl-1H-benzimidazole 95%
6-Chloro-2-ethyl-1H-benzimidazole
Formel: C₉H₉ClN₂
Molecular Weight: 180.634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18800923
CAS-Nummer: 34569-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR