Boc-Cys(stbu)-OH

Lieferant: Apollo Scientific

Synonyme: (R)-2-((tert-Butoxycarbonyl)amino)-3-(tert-butyldisulphanyl)propanoic acid

OR932786-1G OR932786-5G
APOSOR932786-1GEA 94.4 EUR
APOSOR932786-1G APOSOR932786-5G
Boc-Cys(stbu)-OH
Boc-Cys(stbu)-OH
Formel: C₁₂H₂₃NO₄S₂
Schmelzpunkt: 110…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084687
CAS-Nummer: 30044-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR