2,2-Dimethyl-piperazine dihydrochloride 95%

Lieferant: Apollo Scientific

OR932791-1G
APOSOR932791-1GEA 364 EUR
APOSOR932791-1G
2,2-Dimethyl-piperazine dihydrochloride 95%
2,2-Dimethyl-piperazine dihydrochloride
Formel: C₆H₁₄N₂·2ClH
Molecular Weight: 187,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08752613
CAS-Nummer: 128427-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR