2-Amino-4,5-dimethoxybenzonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyano-4,5-dimethoxyaniline

OR9328-5G
APOSOR9328-5GEA 65.4 EUR
APOSOR9328-5G
2-Amino-4,5-dimethoxybenzonitril
2-Amino-4,5-dimethoxybenzonitril
Formel: C₉H₁₀N₂O₂
Molecular Weight: 178,19 g/mol
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00058682
CAS-Nummer: 26961-27-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR