4-Chloro-6-methylnicotinic acid 95%

Lieferant: Apollo Scientific

OR932816-10G OR932816-1G OR932816-5G
APOSOR932816-10GEA 330 EUR
APOSOR932816-10G APOSOR932816-1G APOSOR932816-5G
4-Chloro-6-methylnicotinic acid 95%
4-Chloro-6-methylnicotinic acid
Formel: C₇H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11520850
CAS-Nummer: 1060805-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR