2-Isopropylpyrimidine-5-carboxylic acid 98%
Lieferant: Apollo Scientific
OR932820-1G
APOSOR932820-1GEA
328
EUR
APOSOR932820-1G
2-Isopropylpyrimidine-5-carboxylic acid 98%
2-Isopropylpyrimidine-5-carboxylic acid
Formel:
C₈H₁₀N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09054857 CAS-Nummer: 927803-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...