6,7-Dihydroisoquinolin-8(5H)-one hydrochloride 95%
Lieferant: Apollo Scientific
OR932821-1G
APOSOR932821-1GEA
106
EUR
APOSOR932821-1G
6,7-Dihydroisoquinolin-8(5H)-one hydrochloride 95%
6,7-Dihydroisoquinolin-8(5H)-one hydrochloride
Formel:
C₉H₉NO·ClH Molecular Weight: 183,635 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18432614 CAS-Nummer: 135311-97-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...