Benzofuran-6-ol 95%

Lieferant: Apollo Scientific

OR932830-1G OR932830-250MG
APOSOR932830-1GEA 432 EUR
APOSOR932830-1G APOSOR932830-250MG
Benzofuran-6-ol 95%
Benzofuran-6-ol
Formel: C₈H₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08669481
CAS-Nummer: 13196-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR