1,1-Diethylharnstoff 97%

Lieferant: Apollo Scientific

OR932849-25G OR932849-5G
APOSOR932849-25GEA 270 EUR
APOSOR932849-25G APOSOR932849-5G
1,1-Diethylharnstoff 97%
1,1-Diethylharnstoff
Formel: C₅H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00007960
CAS-Nummer: 634-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR