Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%

Lieferant: Apollo Scientific

Synonyme: TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine

OR932877-1G OR932877-250MG OR932877-5G
APOSOR932877-1GEA 157 EUR
APOSOR932877-1G APOSOR932877-250MG APOSOR932877-5G
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Formel: C₃₀H₃₀N₁₀
Molecular Weight: 530.64 g/mol
Schmelzpunkt: 132…143 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09265124
CAS-Nummer: 510758-28-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR