tert-Butyl glycolate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-hydroxyacetate

OR932881-100G
APOSOR932881-100GEA 172 EUR
APOSOR932881-100G
tert-Butyl glycolate 95%
tert-Butyl glycolate
Formel: C₆H₁₂O₃
Lagertemperatur: Kühlschrank
MDL: MFCD03452739
CAS-Nummer: 50595-15-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR