(2-Chloro-thiazol-5-ylmethyl)-ethyl-amine 95%

Lieferant: Apollo Scientific

Synonyme: N-((2-Chlorothiazol-5-yl)methyl)ethanamine

OR932883-1G
APOSOR932883-1GEA 393 EUR
APOSOR932883-1G
(2-Chloro-thiazol-5-ylmethyl)-ethyl-amine 95%
(2-Chloro-thiazol-5-ylmethyl)-ethyl-amine
Formel: C₆H₉ClN₂S
Molecular Weight: 176.667 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18837199
CAS-Nummer: 120740-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR