(R)-4-Cyano-3-hydrobutyric acid ethyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-cyanobutanoate

OR932907-1G
APOSOR932907-1GEA 163 EUR
APOSOR932907-1G
(R)-4-Cyano-3-hydrobutyric acid ethyl ester 95%
(R)-4-Cyano-3-hydrobutyric acid ethyl ester
Formel: C₇H₁₁NO₂
Molecular Weight: 141,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040664
CAS-Nummer: 10444-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR