Methyl-3-amino-2,2-dimethylpropanoate hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-amino-2,2-dimethylpropanoate hydrochloride
OR932952-1G
APOSOR932952-1GEA
58
EUR
APOSOR932952-1G
Methyl-3-amino-2,2-dimethylpropanoate hydrochloride 95%
Methyl-3-amino-2,2-dimethylpropanoate hydrochloride
Formel:
C₆H₁₄ClNO₂ Molecular Weight: 167,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10758092 CAS-Nummer: 177269-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...