2-P-Tolyloxymethyl-oxirane 95%
Lieferant: Apollo Scientific
Synonyme:
2-((p-Tolyloxy)methyl)oxirane
OR932992-1G
APOSOR932992-1GEA
120
EUR
APOSOR932992-1G
2-P-Tolyloxymethyl-oxirane 95%
2-P-Tolyloxymethyl-oxirane
Formel:
C₁₀H₁₂O₂ Molecular Weight: 164,201 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046991 CAS-Nummer: 2186-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...