2,4,4-Trimethylcyclopentanon 95%

Lieferant: Apollo Scientific

OR932999-5G
APOSOR932999-5GEA 319 EUR
APOSOR932999-5G
2,4,4-Trimethylcyclopentanon 95%
2,4,4-Trimethylcyclopentanon
Formel: C₈H₁₄O
Molecular Weight: 126.2 g/mol
Schmelzpunkt: -25.6
Lagertemperatur: Raumtemperatur
MDL: MFCD00019313
CAS-Nummer: 4694-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR