4,5,6,7-Tetrahydro-1,2-benzoxazol-3-amine 98%

Lieferant: Apollo Scientific

OR933001-1G
APOSOR933001-1GEA 364 EUR
APOSOR933001-1G
4,5,6,7-Tetrahydro-1,2-benzoxazol-3-amine 98%
4,5,6,7-Tetrahydro-1,2-benzoxazol-3-amine
Formel: C₇H₁₀N₂O
Molecular Weight: 138,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19205004
CAS-Nummer: 1004-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR