5-Methyl-1H-pyrrole-3-carboxylic acid methyl ester 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-methyl-1H-pyrrole-3-carboxylate

OR933012-1G
APOSOR933012-1GEA 372 EUR
APOSOR933012-1G
5-Methyl-1H-pyrrole-3-carboxylic acid methyl ester 95%
5-Methyl-1H-pyrrole-3-carboxylic acid methyl ester
Formel: C₇H₉NO₂
Molecular Weight: 139,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00968364
CAS-Nummer: 40611-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR