2-Methylbenzoyl cyanide 95%

Lieferant: Apollo Scientific

OR933121-1G
APOSOR933121-1GEA 603 EUR
APOSOR933121-1G
2-Methylbenzoyl cyanide 95%
2-Methylbenzoyl cyanide
Formel: C₉H₇NO
Molecular Weight: 145,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658474
CAS-Nummer: 5955-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR