1-Formamido-1-cyclohexene 95%

Lieferant: Apollo Scientific

OR933130-1G
APOSOR933130-1GEA 400 EUR
APOSOR933130-1G
1-Formamido-1-cyclohexene 95%
1-Formamido-1-cyclohexene
Formel: C₇H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD01863768
CAS-Nummer: 40652-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR